missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[3-(Benzyloxy)phenyl]propionic acid, 96%, Thermo Scientific Chemicals
31.00 € - 279.00 €
Chemical Identifiers
CAS | 57668-34-5 |
---|---|
Molecular Formula | C16H16O3 |
Molecular Weight (g/mol) | 256.301 |
MDL Number | MFCD09258905 |
InChI Key | HVXOYUOKPLHWNU-UHFFFAOYSA-N |
Synonym | 3-3-benzyloxy phenyl propanoic acid, benzenepropanoic acid, 3-phenylmethoxy, 3-3-benzyloxy-phenyl-propanoic acid, 3-3-benzyloxy phenyl propionic acid, 3-3-phenylmethyl oxy phenyl propanoic acid |
PubChem CID | 21938504 |
IUPAC Name | 3-(3-phenylmethoxyphenyl)propanoic acid |
SMILES | C1=CC=C(C=C1)COC2=CC=CC(=C2)CCC(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15446787
|
Thermo Scientific Alfa Aesar
H33899.MD |
250 mg | |||||||
15426787
|
Thermo Scientific Alfa Aesar
H33899.03 |
1 g |
86.40 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15436787
|
Thermo Scientific Alfa Aesar
H33899.06 |
5 g |
279.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
57668-34-5 | |
256.301 | |
HVXOYUOKPLHWNU-UHFFFAOYSA-N | |
21938504 | |
C1=CC=C(C=C1)COC2=CC=CC(=C2)CCC(=O)O |
C16H16O3 | |
MFCD09258905 | |
3-3-benzyloxy phenyl propanoic acid, benzenepropanoic acid, 3-phenylmethoxy, 3-3-benzyloxy-phenyl-propanoic acid, 3-3-benzyloxy phenyl propionic acid, 3-3-phenylmethyl oxy phenyl propanoic acid | |
3-(3-phenylmethoxyphenyl)propanoic acid |
Specifications
57668-34-5 | |
C16H16O3 | |
3-3-benzyloxy phenyl propanoic acid, benzenepropanoic acid, 3-phenylmethoxy, 3-3-benzyloxy-phenyl-propanoic acid, 3-3-benzyloxy phenyl propionic acid, 3-3-phenylmethyl oxy phenyl propanoic acid | |
C1=CC=C(C=C1)COC2=CC=CC(=C2)CCC(=O)O | |
256.301 | |
256.3 | |
3-[3-(Benzyloxy)phenyl]propionic acid |
83°C to 85°C | |
MFCD09258905 | |
HVXOYUOKPLHWNU-UHFFFAOYSA-N | |
3-(3-phenylmethoxyphenyl)propanoic acid | |
21938504 | |
96% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only