missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Pyrrolidinyl)piperidine, 94%, Thermo Scientific™

Product Code. 15453357
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 858262-19-8
Molecular Formula C9H18N2
MDL Number MFCD08277606
Quantity 100 mg
UN Number UN3259
Sensitivity Air Sensitive
Synonym 3-2-pyrrolidinyl piperidine, 3-pyrrolidin-2-yl-piperidine, 3-pyrrolidin-2-yl piperidine, 3-2-pyrrolidino piperidine, 2-3-piperidinyl pyrrolidine, piperidine,3-2-pyrrolidinyl
InChI Key QLTZNGNUWYPOSS-UHFFFAOYSA-N
SMILES C1CC(CNC1)C2CCCN2
IUPAC Name 3-pyrrolidin-2-ylpiperidine
Molecular Weight (g/mol) 154.257
PubChem CID 458601
Formula Weight 154.26
Percent Purity 94%
Chemical Name or Material 3-(2-Pyrrolidinyl)piperidine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.