missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methylphenoxy)pyrrolidine, 95%, Thermo Scientific Chemicals

75.30 € - 263.00 €

Chemical Identifiers

CAS 46196-54-7
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD08060984
InChI Key MIVMFYNEGPHLSB-UHFFFAOYSA-N
Synonym 3-2-methylphenoxy pyrrolidine, 3-o-tolyloxy pyrrolidine, 3-o-tolyloxy-pyrrolidine, acmc-1bmts, 3-o-tolyloxy ;pyrrolidine
PubChem CID 18337163
IUPAC Name 3-(2-methylphenoxy)pyrrolidine
SMILES CC1=CC=CC=C1OC2CCNC2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11494750
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H50784.MD
250 mg
75.30 €
250mg
Please to purchase this item. Need a web account? Register with us today!
11404760
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H50784.03
1 g
263.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

46196-54-7
177.247
MIVMFYNEGPHLSB-UHFFFAOYSA-N
18337163
CC1=CC=CC=C1OC2CCNC2
C11H15NO
MFCD08060984
3-2-methylphenoxy pyrrolidine, 3-o-tolyloxy pyrrolidine, 3-o-tolyloxy-pyrrolidine, acmc-1bmts, 3-o-tolyloxy ;pyrrolidine
3-(2-methylphenoxy)pyrrolidine

Specifications

46196-54-7
MFCD08060984
MIVMFYNEGPHLSB-UHFFFAOYSA-N
3-(2-methylphenoxy)pyrrolidine
18337163
95%
C11H15NO
3-2-methylphenoxy pyrrolidine, 3-o-tolyloxy pyrrolidine, 3-o-tolyloxy-pyrrolidine, acmc-1bmts, 3-o-tolyloxy ;pyrrolidine
CC1=CC=CC=C1OC2CCNC2
177.247
177.25
3-(2-Methylphenoxy)pyrrolidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only