missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methyl-1,3-dioxolan-2-yl)benzeneboronic acid, 97%, Thermo Scientific™

57.60 € - 181.00 €

Chemical Identifiers

CAS 850568-50-2
Molecular Formula C10H13BO4
Molecular Weight (g/mol) 208.02
MDL Number MFCD04973736
InChI Key NSDRSKQWUQJUAE-UHFFFAOYSA-N
Synonym 3-2-methyl-1,3-dioxolan-2-yl phenylboronic acid, 3-2-methyl-1,3-dioxolan-2-yl phenyl boronic acid, 2-3-boronobenzene-2-methyl-1,3-dioxolane, 2-3-boronophenyl-2-methyl-1,3-dioxolane, 3-2-methyl-1,3-dioxolan-2-yl benzeneboronic acid, 3-boronoacetophenone ethylene acetal, boronic acid,b-3-2-methyl-1,3-dioxolan-2-yl phenyl
PubChem CID 3674880
IUPAC Name [3-(2-methyl-1,3-dioxolan-2-yl)phenyl]boronic acid
SMILES CC1(OCCO1)C1=CC=CC(=C1)B(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15410556
View Documents
Thermo Scientific Alfa Aesar
H53286.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15400556
View Documents
Thermo Scientific Alfa Aesar
H53286.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

850568-50-2
208.02
NSDRSKQWUQJUAE-UHFFFAOYSA-N
3674880
CC1(OCCO1)C1=CC=CC(=C1)B(O)O
C10H13BO4
MFCD04973736
3-2-methyl-1,3-dioxolan-2-yl phenylboronic acid, 3-2-methyl-1,3-dioxolan-2-yl phenyl boronic acid, 2-3-boronobenzene-2-methyl-1,3-dioxolane, 2-3-boronophenyl-2-methyl-1,3-dioxolane, 3-2-methyl-1,3-dioxolan-2-yl benzeneboronic acid, 3-boronoacetophenone ethylene acetal, boronic acid,b-3-2-methyl-1,3-dioxolan-2-yl phenyl
[3-(2-methyl-1,3-dioxolan-2-yl)phenyl]boronic acid

Specifications

850568-50-2
C10H13BO4
3-2-methyl-1,3-dioxolan-2-yl phenylboronic acid, 3-2-methyl-1,3-dioxolan-2-yl phenyl boronic acid, 2-3-boronobenzene-2-methyl-1,3-dioxolane, 2-3-boronophenyl-2-methyl-1,3-dioxolane, 3-2-methyl-1,3-dioxolan-2-yl benzeneboronic acid, 3-boronoacetophenone ethylene acetal, boronic acid,b-3-2-methyl-1,3-dioxolan-2-yl phenyl
CC1(OCCO1)C1=CC=CC(=C1)B(O)O
208.02
208.02
3-(2-Methyl-1,3-dioxolan-2-yl)benzeneboronic acid
100°C to 104°C
MFCD04973736
NSDRSKQWUQJUAE-UHFFFAOYSA-N
[3-(2-methyl-1,3-dioxolan-2-yl)phenyl]boronic acid
3674880
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications