missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Aminoethylamino)propyltrimethoxysilane, 96%, Thermo Scientific Chemicals

Product Code. 11368925
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3-(2-Aminoethylamino) propyltrimethoxysilane acts as a silane coupling agent. It is useful for chemical research. It is also employed as an adhesives and sealant chemical. Further, it is used as a paint additive and coating additive. In addition to this, it plays an important role in the production of petroleum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(2-Aminoethylamino) propyltrimethoxysilane acts as a silane coupling agent. It is useful for chemical research. It is also employed as an adhesives and sealant chemical. Further, it is used as a paint additive and coating additive. In addition to this, it plays an important role in the production of petroleum.

Solubility
Miscible with toluene.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1760-24-3
Molecular Formula C8H22N2O3Si
Molecular Weight (g/mol) 222.36
MDL Number MFCD00008173
InChI Key PHQOGHDTIVQXHL-UHFFFAOYSA-N
Synonym n-3-trimethoxysilyl propyl ethylenediamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 3-2-aminoethylamino propyltrimethoxysilane, en-aptas, silicone a-1120, prosil 3128, aas-m, n1-3-trimethoxysilyl propyl ethane-1,2-diamine, unii-28zcs5ga8g, 3-2-aminoethyl aminopropyl trimethoxysilane
PubChem CID 15659
IUPAC Name N'-(3-trimethoxysilylpropyl)ethane-1,2-diamine
SMILES CO[Si](CCCNCCN)(OC)OC

Specifications

Boiling Point 261°C to 263°C
Quantity 100 g
UN Number UN2735
Beilstein 636230
Flash Point 136°C (276°F)
Solubility Information Miscible with toluene.
Formula Weight 222.36
Percent Purity 96%
Odor Ammonia-like
Refractive Index 1.4435
Sensitivity Air and moisture sensitive
Density 1.01
Chemical Name or Material 3-(2-Aminoethylamino)propyltrimethoxysilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'