missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1-Naphthyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals
27.00 € - 283.00 €
Chemical Identifiers
CAS | 881913-20-8 |
---|---|
Molecular Formula | C16H13BO2 |
Molecular Weight (g/mol) | 248.088 |
MDL Number | MFCD11045054 |
InChI Key | HFXYUCJLQZCNPD-UHFFFAOYSA-N |
Synonym | 3-naphthalen-1-yl phenyl boronic acid, 3-naphthalen-1-yl phenylboronic acid, 3-1-naphthyl phenylboronic acid, 3-1-naphthyl benzeneboronic acid, boronic acid, 3-1-naphthalenyl phenyl, 3-1-naphthyl phenyl boronic acid, 3-1-naphthalenyl phenyl-boronic acid |
PubChem CID | 58881077 |
IUPAC Name | (3-naphthalen-1-ylphenyl)boronic acid |
SMILES | B(C1=CC(=CC=C1)C2=CC=CC3=CC=CC=C32)(O)O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
881913-20-8 | |
248.088 | |
HFXYUCJLQZCNPD-UHFFFAOYSA-N | |
58881077 | |
B(C1=CC(=CC=C1)C2=CC=CC3=CC=CC=C32)(O)O |
C16H13BO2 | |
MFCD11045054 | |
3-naphthalen-1-yl phenyl boronic acid, 3-naphthalen-1-yl phenylboronic acid, 3-1-naphthyl phenylboronic acid, 3-1-naphthyl benzeneboronic acid, boronic acid, 3-1-naphthalenyl phenyl, 3-1-naphthyl phenyl boronic acid, 3-1-naphthalenyl phenyl-boronic acid | |
(3-naphthalen-1-ylphenyl)boronic acid |
Specifications
881913-20-8 | |
C16H13BO2 | |
18694809 | |
HFXYUCJLQZCNPD-UHFFFAOYSA-N | |
(3-naphthalen-1-ylphenyl)boronic acid | |
58881077 | |
98% | |
3-(1-Naphthyl)benzeneboronic acid |
White | |
MFCD11045054 | |
3-naphthalen-1-yl phenyl boronic acid, 3-naphthalen-1-yl phenylboronic acid, 3-1-naphthyl phenylboronic acid, 3-1-naphthyl benzeneboronic acid, boronic acid, 3-1-naphthalenyl phenyl, 3-1-naphthyl phenyl boronic acid, 3-1-naphthalenyl phenyl-boronic acid | |
B(C1=CC(=CC=C1)C2=CC=CC3=CC=CC=C32)(O)O | |
248.088 | |
248.09 | |
Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only