missing translation for 'onlineSavingsMsg'
Learn More

Sharpsafe™ 24L Infectious Sharps for Incineration Waste Container

The generous size of the 24L model means it is particularly suited for areas generating large amounts of contaminated waste.

Brand:  Sharpsafe™ 41202411

 View more versions of this product

Product Code. 15635697

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Wide horizontal aperture ensures easy disposal of large items and allows proportionally greater fill than similar vertical feed type openings
  • Large volume provides enhanced cost effectiveness in high usage areas
  • A semi-translucent lid and visible fill line help ensure user safety by allowing clear indication of when the fill line is reached
  • Pivot flap ensures safe disposal
  • In-mould label offers permanent audit trail which cannot be removed
  • Temporary and Final Closure feature
  • Simple 4-click assembly

Specifications

Specifications

24 L
Orange Semi-Translucent
Closure
UN 3291, BS EN ISO 23907:2012
335 x 230 x 450 mm
Disposal of sharp, contaminated waste
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.