missing translation for 'onlineSavingsMsg'
Learn More

HUBER Insulated Stainless Steel Bath 212B, 12 L

Insulated bath

Brand:  HUBER 6684

Product Code. 10692415

  • 1185.00 € / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Insulated stainless steel bath 212B, 12L
Specifications

Specifications

12 L
200°C
212 B
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers