Learn More
Ohaus™ 2 x Centrifuge Tube Adapters, Frontier™ R-A6x250/8MI Fixed Rotors
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ R-A6x250/8MI Fixed Rotors
Brand: Ohaus™ 30304373
This item is not returnable.
View return policy
Description
- Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
- Suitable for use in Frontier™ R-A6x250/8MI Fixed Rotors
Specifications
RB | |
29 mm | |
Adapter, 1x50ml D29mm RB, 2/pk | |
2 |
1 x 50 mL (29 mm dia.) | |
R-A6x250/8MI Fixed Rotors | |
1 |