Learn More
Ohaus™ 2 x Centrifuge Racks, Frontier™ R-S4x100/5MIS Swing Out Rotors
Fit inside a swing-out bucket to facilitate centrifugation of tubes of custom size/number.
Brand: Ohaus™ 30314880
This item is not returnable.
View return policy
Description
- Allows you to centrifuge tubes of custom size or number
- Suitable for use in buckets for Frontier™ R-S4x100/5MIS Swing Out Rotors
Specifications
4 x 10 mL | |
R-S4x100/5MIS Swing Out Rotors | |
4 |
17 | |
Rack, 4x10ml D17mm Vac, 2/pk | |
2 |