Learn More
Ohaus™ 2 x Centrifuge Racks, Frontier™ R-S4x100/5MIS Swing Out Rotors
Fit inside a swing-out bucket to facilitate centrifugation of tubes of custom size/number.
Brand:  Ohaus™ 30314876
 View more versions of this product
Product Code. 15826712
  • 180.00 € / Pack of 2
Explore more special offers
Description
Description
  • Allows you to centrifuge tubes of custom size or number
  • Suitable for use in buckets for Frontier™ R-S4x100/5MIS Swing Out Rotors