Learn More
Ohaus™ 2 x Centrifuge Rack Adapters, Frontier™ R-S4x250/4MI Swing Out Rotors
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ R-S4x250/4MI Swing Out Rotors
Brand:  Ohaus™ 30314903
185.39 EUR valid until 2024-11-30
Use promo code "21681" to get your promotional price.
 View more versions of this product
Product Code. 15896732
  • 277.00 € / Pack of 2
Explore more special offers
Description
Description
  • Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
  • Suitable for use in Frontier™ R-S4x250/4MI Swing Out Rotors
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FB
62 mm
Rack, 1x250ml D62mm FB, 2/pk
2
1 x 250 mL
R-S4x250/4MI Swing Out Rotors
1