Learn More
Ohaus™ 2 x Centrifuge Rack Adapters, Frontier™ R-S4x250/4MI Swing Out Rotors
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ R-S4x250/4MI Swing Out Rotors
Brand: Ohaus™ 30314909
This item is not returnable.
View return policy
Description
- Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
- Suitable for use in Frontier™ R-S4x250/4MI Swing Out Rotors
Specifications
Rim | |
29 mm | |
Rack, 1x50ml D29mm Rim, 2/pk, V3 | |
2 |
1 x 50 mL (29 mm dia. Rim) | |
R-S4x250/4MI Swing Out Rotors | |
1 |