Learn More
Ohaus™ 2 x Centrifuge Rack Adapters, Frontier™ R-S4x250/4MI Swing Out Rotors
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ R-S4x250/4MI Swing Out Rotors
Brand: Ohaus™ 30314906
This item is not returnable.
View return policy
Description
- Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
- Suitable for use in Frontier™ R-S4x250/4MI Swing Out Rotors
Specifications
FA | |
29 mm | |
Rack, 3x50ml D29mm FA, 2/pk | |
2 |
3 x 50 mL (29 mm dia. FA) | |
R-S4x250/4MI Swing Out Rotors | |
3 |