Learn More
Ohaus™ 2 x Centrifuge Rack Adapters, Frontier™ R-S4x100/4MI Swing Out Rotors
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ R-S4x100/4MI Swing Out Rotors
Brand:  Ohaus™ 30314844
191.50 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15816682
  • 200.00 € / Pack of 2
Explore more special offers
Description
Description
  • Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
  • Suitable for use in Frontier™ R-S4x100/4MI Swing Out Rotors
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RB
34 mm
Rack, 1x50ml D34mm RB, 2/pk
2
1 x 50 mL (34 mm dia.)
R-S4x100/4MI Swing Out Rotors
1