Learn More
2-Vinylpyridine, 97%, stabilized, Thermo Scientific Chemicals
31.26 € - 189.95 €
Chemical Identifiers
CAS | 100-69-6 |
---|---|
Molecular Formula | C7H7N |
Molecular Weight (g/mol) | 105.14 |
MDL Number | MFCD00006355 |
InChI Key | KGIGUEBEKRSTEW-UHFFFAOYSA-N |
Synonym | 2-vinylpyridine, pyridine, 2-ethenyl, pyridine, 2-vinyl, pyridine, ethenyl, vinylpyridine, 2-vinyl pyridine, alpha-vinylpyridine, vinyl pyridine, unii-dt4uv4nnkx, ccris 5238 |
PubChem CID | 7521 |
IUPAC Name | 2-ethenylpyridine |
SMILES | C=CC1=CC=CC=N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10754461
|
Thermo Scientific Acros
158000050 |
5 mL | Glass bottle |
31.26 €
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10295530
|
Thermo Scientific Acros
158002500 |
250 mL | Glass bottle |
62.71 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10589633
|
Thermo Scientific Acros
158000010 |
1 L | Glass bottle |
189.95 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100-69-6 | |
105.14 | |
KGIGUEBEKRSTEW-UHFFFAOYSA-N | |
7521 | |
C=CC1=CC=CC=N1 |
C7H7N | |
MFCD00006355 | |
2-vinylpyridine, pyridine, 2-ethenyl, pyridine, 2-vinyl, pyridine, ethenyl, vinylpyridine, 2-vinyl pyridine, alpha-vinylpyridine, vinyl pyridine, unii-dt4uv4nnkx, ccris 5238 | |
2-ethenylpyridine |
Specifications
100-69-6 | |
100.0 | |
Brown | |
159°C | |
Authentic | |
C7H7N | |
1.5490 to 1.551 | |
20, 256 | |
2-vinylpyridine, pyridine, 2-ethenyl, pyridine, 2-vinyl, pyridine, ethenyl, vinylpyridine, 2-vinyl pyridine, alpha-vinylpyridine, vinyl pyridine, unii-dt4uv4nnkx, ccris 5238 | |
KGIGUEBEKRSTEW-UHFFFAOYSA-N | |
2-ethenylpyridine | |
105.14 | |
1.2 mPa.s (20°C) | |
97% | |
2-Vinylpyridine, 97% |
96.0 | |
-50°C | |
0.9983g/mL | |
47°C | |
96% min. (GC) | |
(C5H4N)CH=CH2 | |
MFCD00006355 | |
0.9983 | |
Solubility in water: 2.5g/100 mL in water (20°C). Other solubilities: soluble in dilute acids,soluble in hydrocarbons,alcohol,ketones and,esters,soluble in methanol,ethanol,dioxane | |
C=CC1=CC=CC=N1 | |
0.1% tert.-butylcatechol | |
7521 | |
105.14 | |
Liquid |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vap
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-879-8
missing translation for 'rtecsNumber' : UU1040000
missing translation for 'tsca' : TSCA
- 2-Vinylpyridine, stabilized
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- H311-Toxic in contact with skin.
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.