missing translation for 'onlineSavingsMsg'
Learn More

2-Valerylthiophene, 97%, Thermo Scientific™

Product Code. 11401210
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53119-25-8
Molecular Formula C9H12OS
Molecular Weight (g/mol) 168.254
MDL Number MFCD00053149
InChI Key KJMPRCVRMAHTBR-UHFFFAOYSA-N
Synonym 1-thien-2-ylpentan-1-one, 1-2-thienyl pentan-1-one, 1-thiophen-2-yl pentan-1-one, 1-pentanone, 1-2-thienyl, unii-x71z1k8ray, 2-valerylthiophene, x71z1k8ray, 2-pentanoylthiophene, butyl 2-thienyl ketone, acmc-20aors
PubChem CID 574100
IUPAC Name 1-thiophen-2-ylpentan-1-one
SMILES CCCCC(=O)C1=CC=CS1

Specifications

Refractive Index 1.5358
Quantity 1 g
Formula Weight 168.25
Percent Purity 97%
Chemical Name or Material 2-Valerylthiophene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'