missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)pyridine-5-boronic acid, 98%, Thermo Scientific Chemicals

Product Code. 15438706
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868662-36-6
Molecular Formula C6H5BF3NO2
Molecular Weight (g/mol) 190.916
MDL Number MFCD07375381
InChI Key BNTIPMNMTIAWIW-UHFFFAOYSA-N
Synonym 2-trifluoromethyl pyridine-5-boronic acid, 6-trifluoromethyl pyridin-3-yl boronic acid, 2-trifluoromethyl-5-pyridineboric acid, 2-trifluoromethyl pyridin-5-ylboronic acid, 6-trifluoromethyl pyridine-3-boronic acid, 6-trifluoromethyl pyridin-3-ylboronic acid, 2-trifluoromethylpyridine-5-boronic acid, 2-trifluoromethyl-5-pyridineboronic acid, 6-trifluoromethyl-3-pyridylboronic acid
PubChem CID 25134278
IUPAC Name [6-(trifluoromethyl)pyridin-3-yl]boronic acid
SMILES B(C1=CN=C(C=C1)C(F)(F)F)(O)O

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 190.92
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-(Trifluoromethyl)pyridine-5-boronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.