missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-2-(Trifluoromethyl)piperidine, 97%, Thermo Scientific™
157.00 € - 528.00 €
Chemical Identifiers
CAS | 154630-93-0 |
---|---|
Molecular Formula | C6H10F3N |
Molecular Weight (g/mol) | 153.15 |
InChI Key | NAXDEFXCCITWEU-UHFFFAOYSA-N |
Synonym | 2-trifluoromethyl piperidine, 2-trifluoromethyl-piperidine, -2-trifluoromethyl piperidine, piperidine, 2-trifluoromethyl, +/--2-trifluoromethyl piperidine |
PubChem CID | 11263608 |
IUPAC Name | 2-(trifluoromethyl)piperidine |
SMILES | C1CCNC(C1)C(F)(F)F |
Description
Chemical Identifiers
154630-93-0 | |
153.15 | |
2-trifluoromethyl piperidine, 2-trifluoromethyl-piperidine, -2-trifluoromethyl piperidine, piperidine, 2-trifluoromethyl, +/--2-trifluoromethyl piperidine | |
2-(trifluoromethyl)piperidine |
C6H10F3N | |
NAXDEFXCCITWEU-UHFFFAOYSA-N | |
11263608 | |
C1CCNC(C1)C(F)(F)F |
Specifications
154630-93-0 | |
25°C | |
96% min. (GC) | |
2-trifluoromethyl piperidine, 2-trifluoromethyl-piperidine, -2-trifluoromethyl piperidine, piperidine, 2-trifluoromethyl, +/--2-trifluoromethyl piperidine | |
C1CCNC(C1)C(F)(F)F | |
153.15 | |
153.15 | |
(±)-2-(Trifluoromethyl)piperidine |
64.0°C (182.0 mmHg) | |
Authentic | |
C6H10F3N | |
NAXDEFXCCITWEU-UHFFFAOYSA-N | |
2-(trifluoromethyl)piperidine | |
11263608 | |
97% |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- (±)-2-(Trifluoromethyl)piperidine, 97%
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title