missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)phenol, 97%, Thermo Scientific™

37.80 € - 151.00 €

Chemical Identifiers

CAS 32858-93-8
Molecular Formula C7H5F3O2
Molecular Weight (g/mol) 178.11
MDL Number MFCD00236324
InChI Key GQWMNVOVQZIPJC-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy phenol, 2-trifluorormethoxy phenol, 2-hydroxyphenyl trifluoromethyl ether, o-trifluoromethoxyphenol, 2-trifluormethoxy phenol, trifluoromethoxyphenol, phenol, 2-trifluoromethoxy, trifluoromethoxy phenol, acmc-1cmde
PubChem CID 2777299
IUPAC Name 2-(trifluoromethoxy)phenol
SMILES C1=CC=C(C(=C1)O)OC(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11434287
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B20376.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11444287
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B20376.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

32858-93-8
178.11
GQWMNVOVQZIPJC-UHFFFAOYSA-N
2777299
C1=CC=C(C(=C1)O)OC(F)(F)F
C7H5F3O2
MFCD00236324
2-trifluoromethoxy phenol, 2-trifluorormethoxy phenol, 2-hydroxyphenyl trifluoromethyl ether, o-trifluoromethoxyphenol, 2-trifluormethoxy phenol, trifluoromethoxyphenol, phenol, 2-trifluoromethoxy, trifluoromethoxy phenol, acmc-1cmde
2-(trifluoromethoxy)phenol

Specifications

32858-93-8
69°C to 71°C (60 mmHg)
1.443
1869013
GQWMNVOVQZIPJC-UHFFFAOYSA-N
2-(trifluoromethoxy)phenol
2777299
97%
1.332
C7H5F3O2
MFCD00236324
2-trifluoromethoxy phenol, 2-trifluorormethoxy phenol, 2-hydroxyphenyl trifluoromethyl ether, o-trifluoromethoxyphenol, 2-trifluormethoxy phenol, trifluoromethoxyphenol, phenol, 2-trifluoromethoxy, trifluoromethoxy phenol, acmc-1cmde
C1=CC=C(C(=C1)O)OC(F)(F)F
178.11
178.11
2-(Trifluoromethoxy)phenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates