missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrrolidin-1-yl-1,3-thiazole-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10103414
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 900015-48-7
Molecular Formula C8H10N2OS
Molecular Weight (g/mol) 182.241
MDL Number MFCD07021330
InChI Key CFFWKLLEEVWNLE-UHFFFAOYSA-N
Synonym 2-1-pyrrolidinyl-1,3-thiazole-5-carbaldehyde, 2-pyrrolidin-1-yl-1,3-thiazole-5-carbaldehyde, 2-pyrrolidinyl-1,3-thiazole-5-carbaldehyde, 2-pyrrolidin-1-yl thiazole-5-carbaldehyde
PubChem CID 2767663
IUPAC Name 2-pyrrolidin-1-yl-1,3-thiazole-5-carbaldehyde
SMILES C1CCN(C1)C2=NC=C(S2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 182.25
Percent Purity 97%
Chemical Name or Material 2-Pyrrolidin-1-yl-1, 3-thiazole-5-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'