Learn More
2-Pyrazinylmethanol, 97%, Thermo Scientific™
46.70 € - 415.00 €
Chemische Identifikatoren
CAS | 6705-33-5 |
---|---|
Molecular Formula | C5H6N2O |
Molecular Weight (g/mol) | 110.12 |
MDL Number | MFCD01693938 |
InChI Key | LFCWHDGQCWJKCG-UHFFFAOYSA-N |
Synonym | 2-pyrazinylmethanol, pyrazin-2-yl methanol, pyrazinemethanol, 2-hydroxymethyl pyrazine, 2-pyrazinemethanol, 2-hydroxymethylpyrazine, pyrazin-2-yl-methanol, unii-c27z7qi77d, pyrazin-2-ylmethan-1-ol |
PubChem CID | 201734 |
SMILES | OCC1=CN=CC=N1 |
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
10722663
|
Thermo Scientific Maybridge
CC03409CB |
250 mg | Amber Glass Bottle |
46.70 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10059081
|
Thermo Scientific Maybridge
CC03409DA |
1 g | Amber Glass Bottle |
121.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10526314
|
Thermo Scientific Maybridge
CC03409DE |
5 g | Amber Glass Bottle |
415.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
Chemische Identifikatoren
6705-33-5 | |
110.12 | |
LFCWHDGQCWJKCG-UHFFFAOYSA-N | |
201734 |
C5H6N2O | |
MFCD01693938 | |
2-pyrazinylmethanol, pyrazin-2-yl methanol, pyrazinemethanol, 2-hydroxymethyl pyrazine, 2-pyrazinemethanol, 2-hydroxymethylpyrazine, pyrazin-2-yl-methanol, unii-c27z7qi77d, pyrazin-2-ylmethan-1-ol | |
OCC1=CN=CC=N1 |
Spezifikation
6705-33-5 | |
100.0 | |
C5H6N2O | |
2-pyrazinylmethanol, pyrazin-2-yl methanol, pyrazinemethanol, 2-hydroxymethyl pyrazine, 2-pyrazinemethanol, 2-hydroxymethylpyrazine, pyrazin-2-yl-methanol, unii-c27z7qi77d, pyrazin-2-ylmethan-1-ol | |
OCC1=CN=CC=N1 | |
110.12 | |
110.12 | |
2-Pyrazinylmethanol |
94.5 | |
95% | |
MFCD01693938 | |
LFCWHDGQCWJKCG-UHFFFAOYSA-N | |
(pyrazin-2-yl)methanol | |
201734 | |
95% |
Sicherheit und Handhabung
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.