missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Pyrazinecarbonyl chloride, Technical Grade, Thermo Scientific™
78.90 € - 439.00 €
Chemical Identifiers
CAS | 19847-10-0 |
---|---|
Molecular Formula | C5H3ClN2O |
Molecular Weight (g/mol) | 142.542 |
InChI Key | TXJKATOSKLUITR-UHFFFAOYSA-N |
Synonym | 2-pyrazinecarbonyl chloride, pyrazinecarbonyl chloride, pyrazine-2-carboxylic acid chloride, 2-pyrazinecarbonylchloride, pyrazine 2-carbonyl chloride, 2-pyrazine carbonyl chloride, pyrazinoyl chloride, acmc-20ann0, 2-pyrazine carboxylic acid chloride, pyrazine-2-carbonyl chloride, technical 1g |
PubChem CID | 88279 |
IUPAC Name | pyrazine-2-carbonyl chloride |
SMILES | C1=CN=C(C=N1)C(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10049081
|
Thermo Scientific Maybridge
CC03402CB |
250 mg | Amber Glass Bottle |
78.90 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10712663
|
Thermo Scientific Maybridge
CC03402DA |
1 g | Amber Glass Bottle |
196.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10662655
|
Thermo Scientific Maybridge
CC03402DE |
5 g | Amber Glass Bottle |
439.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
19847-10-0 | |
142.542 | |
2-pyrazinecarbonyl chloride, pyrazinecarbonyl chloride, pyrazine-2-carboxylic acid chloride, 2-pyrazinecarbonylchloride, pyrazine 2-carbonyl chloride, 2-pyrazine carbonyl chloride, pyrazinoyl chloride, acmc-20ann0, 2-pyrazine carboxylic acid chloride, pyrazine-2-carbonyl chloride, technical 1g | |
pyrazine-2-carbonyl chloride |
C5H3ClN2O | |
TXJKATOSKLUITR-UHFFFAOYSA-N | |
88279 | |
C1=CN=C(C=N1)C(=O)Cl |
Specifications
19847-10-0 | |
2-pyrazinecarbonyl chloride, pyrazinecarbonyl chloride, pyrazine-2-carboxylic acid chloride, 2-pyrazinecarbonylchloride, pyrazine 2-carbonyl chloride, 2-pyrazine carbonyl chloride, pyrazinoyl chloride, acmc-20ann0, 2-pyrazine carboxylic acid chloride, pyrazine-2-carbonyl chloride, technical 1g | |
C1=CN=C(C=N1)C(=O)Cl | |
142.542 | |
142.54 | |
Technical |
C5H3ClN2O | |
TXJKATOSKLUITR-UHFFFAOYSA-N | |
pyrazine-2-carbonyl chloride | |
88279 | |
Tech. | |
2-Pyrazinecarbonyl chloride |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title