missing translation for 'onlineSavingsMsg'
Learn More

2-Propionylthiophene, 98%, Thermo Scientific Chemicals

23.90 € - 210.00 €

Chemical Identifiers

CAS 13679-75-9
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00005446
InChI Key MFPZQZZWAMAHOY-UHFFFAOYSA-N
Synonym 1-2-thienyl-1-propanone, 2-propionylthiophene, 1-thiophen-2-yl propan-1-one, 1-2-thienyl propan-1-one, ethyl 2-thienyl ketone, 2-propanoylthiophene, 1-propanone, 1-2-thienyl, unii-ny658tvp5m, ethyl thienyl ketone, ethyl-2-thienyl ketone
PubChem CID 26179
IUPAC Name 1-thiophen-2-ylpropan-1-one
SMILES CCC(=O)C1=CC=CS1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11327665
View Documents
Thermo Scientific Alfa Aesar
L02318.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11337665
View Documents
Thermo Scientific Alfa Aesar
L02318.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11347665
View Documents
Thermo Scientific Alfa Aesar
L02318.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13679-75-9
140.2
MFPZQZZWAMAHOY-UHFFFAOYSA-N
26179
CCC(=O)C1=CC=CS1
C7H8OS
MFCD00005446
1-2-thienyl-1-propanone, 2-propionylthiophene, 1-thiophen-2-yl propan-1-one, 1-2-thienyl propan-1-one, ethyl 2-thienyl ketone, 2-propanoylthiophene, 1-propanone, 1-2-thienyl, unii-ny658tvp5m, ethyl thienyl ketone, ethyl-2-thienyl ketone
1-thiophen-2-ylpropan-1-one

Specifications

13679-75-9
228°C
C7H8OS
MFCD00005446
1-2-thienyl-1-propanone, 2-propionylthiophene, 1-thiophen-2-yl propan-1-one, 1-2-thienyl propan-1-one, ethyl 2-thienyl ketone, 2-propanoylthiophene, 1-propanone, 1-2-thienyl, unii-ny658tvp5m, ethyl thienyl ketone, ethyl-2-thienyl ketone
CCC(=O)C1=CC=CS1
140.2
140.2
2-Propionylthiophene
1.124
100°C (212°F)
1.553
108934
MFPZQZZWAMAHOY-UHFFFAOYSA-N
1-thiophen-2-ylpropan-1-one
26179
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'einecsNumber' : 237-182-8

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.