missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol CHROMASOLV™, for HPLC, 99.9%, Honeywell Riedel-de Haën™

Product Code. 15634600
Click to view available options
Quantity:
1 L
100 mL
185 L
2 L
2.5 L
2.5 L-PC
4 x 2.5 L
6 x 1 L
72 x 2.5 L

CHROMASOLV™, for HPLC, 99.9%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -89.5°C
Density 0.785g/mL at 25°C
Boiling Point 82°C
Quantity 6 x 1 L
Refractive Index n20/D 1.377
Linear Formula (CH3)2CHOH
Vapor Pressure 44mmHg (25°C)
Beilstein 635639
Formula Weight 60.1g/mol
Percent Purity 99.9%
Grade CHROMASOLV™
Chemical Name or Material 2-Propanol, for HPLC
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Propanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'