missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol, BAKER ANALYZED™ ACS, J.T. Baker™
Click to view available options
1 L
Specifications
Specifications
Melting Point | -89°C |
CAS | 67-63-0 |
Color | Max. 10 (APHA) |
Density | 0.786 g/cm³ (20°C) |
Boiling Point | 82°C (1013 hPa) |
Quantity | 1 L |
Flash Point | 12°C |
Packaging | Glass Bottle |
Assay Percent Range | min. 99.5% |
Residue after Evaporation | max. 0.001% |
Show More |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction