missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol, 99%, BAKER, J.T. Baker™
For laboratory use, for organic synthesis, and for routine research.
98.00 € - 398.00 €
Chemical Identifiers
CAS | 67-63-0 |
---|---|
Molecular Formula | C3H8O |
Molecular Weight (g/mol) | 60.096 |
InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
PubChem CID | 3776 |
ChEBI | CHEBI:17824 |
IUPAC Name | propan-2-ol |
SMILES | CC(C)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10200871
|
JT Baker
8119.2500 |
2.5 L | HDPE |
98.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10721582
|
JT Baker
8119.9025 |
25 L | Steel |
398.00 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
A grade of chemicals for routine research usually controlled by physical criteria and often by assay (by GC).
Chemical Identifiers
67-63-0 | |
60.096 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CHEBI:17824 | |
CC(C)O |
C3H8O | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
3776 | |
propan-2-ol |
Specifications
-88.5°C | |
Colorless | |
Max. 0.002% | |
82.5°C | |
12°C | |
Alcohol | |
Max. 0.005% | |
6.053 kPa at 25°C | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CC(C)O | |
60.096 | |
CHEBI:17824 | |
≥99% | |
2-Propanol |
67-63-0 | |
Max. 0.002% | |
0.781 to 0.783 g/mL at 25°C | |
2.5 - 12 Vol % | |
399°C | |
Liquid | |
C3H8O | |
Max. 0.2% | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol | |
3776 | |
60.1g/mol | |
US FDA / ISO 9000 manufacturing, cGMP, IPEC member |
Safety and Handling
missing translation for 'einecsNumber' : 200-661-7
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title