missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Product Code. 15455325
Click to view available options
Quantity:
50 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Reacts with water.

Notes
Air Sensitive. Store under dry inert gas. Store away from air. Incompatible with oxidizing agents, acid chlorides. Protect from humidity and water. Do not together with acids.
TRUSTED_SUSTAINABILITY

Specifications

CAS 308796-14-7
Density 0.965
Flash Point −17°C (1°F)
Molecular Formula C8H9BrZn
Linear Formula C6H5CH2CH2ZnBr
MDL Number MFCD01311437
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym phenethylzinc bromide, phenethylzinc bromide solution, bromo 2-phenylethyl zinc, phenethylzinc bromide, 0.50 m in thf, phenethylzinc bromide 0.5 m in tetrahydrofuran, phenethylzinc bromide solution, 0.5 m in thf, 2-phenylethylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
Solubility Information Reacts with water.
InChI Key YHVLYRZWQLDARE-UHFFFAOYSA-M
SMILES [CH2-]CC1=CC=CC=C1.[Zn+]Br
IUPAC Name bromozinc(1+);ethylbenzene
Molecular Weight (g/mol) 250.444
PubChem CID 5214896
Formula Weight 250.44
Chemical Name or Material 2-Phenylethylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Phenylethylzinc bromide, 0.5M in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
Supplemental information
  • MIXTURE LIST-Contains : Tetrahydrofuran, 2-Phenylethylzinc bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.