missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenylbenzimidazole-5-sulfonic acid, 98%, Thermo Scientific Chemicals
22.61 € - 227.13 €
Chemical Identifiers
CAS | 27503-81-7 |
---|---|
Molecular Formula | C13H10N2O3S |
Molecular Weight (g/mol) | 274.29 |
MDL Number | MFCD00053007 |
InChI Key | UVCJGUGAGLDPAA-UHFFFAOYSA-N |
Synonym | ensulizole, 2-phenylbenzimidazole-5-sulfonic acid, phenylbenzimidazole sulfonic acid, 2-phenyl-1h-benzo d imidazole-5-sulfonic acid, 2-phenyl-1h-benzo d imidazole-6-sulfonic acid, 1h-benzimidazole-5-sulfonic acid, 2-phenyl, unii-9yq9di1w42, 2-phenyl-1h-benzimidazole-5-sulfonic acid, parsol hs, 2-phenyl-5-benzimidazolesulfonic acid |
PubChem CID | 33919 |
IUPAC Name | 2-phenyl-3H-benzimidazole-5-sulfonic acid |
SMILES | OS(=O)(=O)C1=CC=C2N=C(NC2=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10611833
|
Thermo Scientific Acros
259700050 |
5 g | Glass bottle |
22.61 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10792471
|
Thermo Scientific Acros
259700250 |
25 g | Glass bottle |
40.60 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10259153
|
Thermo Scientific Acros
259702500 |
250 g | Glass bottle |
227.13 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
27503-81-7 | |
274.29 | |
UVCJGUGAGLDPAA-UHFFFAOYSA-N | |
33919 | |
OS(=O)(=O)C1=CC=C2N=C(NC2=C1)C1=CC=CC=C1 |
C13H10N2O3S | |
MFCD00053007 | |
ensulizole, 2-phenylbenzimidazole-5-sulfonic acid, phenylbenzimidazole sulfonic acid, 2-phenyl-1h-benzo d imidazole-5-sulfonic acid, 2-phenyl-1h-benzo d imidazole-6-sulfonic acid, 1h-benzimidazole-5-sulfonic acid, 2-phenyl, unii-9yq9di1w42, 2-phenyl-1h-benzimidazole-5-sulfonic acid, parsol hs, 2-phenyl-5-benzimidazolesulfonic acid | |
2-phenyl-3H-benzimidazole-5-sulfonic acid |
Specifications
27503-81-7 | |
100.0 | |
White | |
98% | |
MFCD00053007 | |
ensulizole, 2-phenylbenzimidazole-5-sulfonic acid, phenylbenzimidazole sulfonic acid, 2-phenyl-1h-benzo d imidazole-5-sulfonic acid, 2-phenyl-1h-benzo d imidazole-6-sulfonic acid, 1h-benzimidazole-5-sulfonic acid, 2-phenyl, unii-9yq9di1w42, 2-phenyl-1h-benzimidazole-5-sulfonic acid, parsol hs, 2-phenyl-5-benzimidazolesulfonic acid | |
UVCJGUGAGLDPAA-UHFFFAOYSA-N | |
2-phenyl-3H-benzimidazole-5-sulfonic acid | |
33919 | |
98% | |
2-Phenylbenzimidazole-5-sulfonic acid |
97.5 | |
>300°C | |
Authentic | |
C13H10N2O3S | |
15, 3647 | |
Solubility in water: soluble | |
OS(=O)(=O)C1=CC=C2N=C(NC2=C1)C1=CC=CC=C1 | |
274.29 | |
274.28 | |
Fine Crystalline Powder |
Safety and Handling
missing translation for 'einecsNumber' : 248-502-
Product Identifier
- 2-Phenylbenzimidazole-5-sulfonic acid
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title