Learn More
2-Phenoxyethyl bromide, 98%, Thermo Scientific Chemicals
44.32 € - 411.05 €
Chemical Identifiers
CAS | 589-10-6 |
---|---|
Molecular Formula | C8H9BrO |
Molecular Weight (g/mol) | 201.063 |
MDL Number | MFCD00000234 |
InChI Key | JJFOBACUIRKUPN-UHFFFAOYSA-N |
Synonym | 2-bromoethoxy benzene, 2-bromoethyl phenyl ether, beta-bromophenetole, 2-phenoxyethylbromide, 2-phenoxyethyl bromide, 1-bromo-2-phenoxyethane, benzene, 2-bromoethoxy, beta-phenoxyethyl bromide, 2-bromoethylphenylether, 1-2-bromoethoxy benzene |
PubChem CID | 68526 |
IUPAC Name | 2-bromoethoxybenzene |
SMILES | C1=CC=C(C=C1)OCCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11444547
|
Thermo Scientific Alfa Aesar
B21087.14 |
25 g |
44.32 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11454547
|
Thermo Scientific Alfa Aesar
B21087.22 |
100 g |
126.63 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11464547
|
Thermo Scientific Alfa Aesar
B21087.36 |
500 g |
411.05 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in the synthesis and antihypertensive activity of 4'-substituted spiro[4H-3,1-benzoxazine-4,4'-piperidin]-2(1H)-ones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the synthesis and antihypertensive activity of 4′-substituted spiro[4H-3,1-benzoxazine-4,4′-piperidin]-2(1H)-ones.
Solubility
Insoluble in water. Solubility in methanol is almost transparent. Soluble in chloroform and ethyl acetate.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
589-10-6 | |
201.063 | |
JJFOBACUIRKUPN-UHFFFAOYSA-N | |
68526 | |
C1=CC=C(C=C1)OCCBr |
C8H9BrO | |
MFCD00000234 | |
2-bromoethoxy benzene, 2-bromoethyl phenyl ether, beta-bromophenetole, 2-phenoxyethylbromide, 2-phenoxyethyl bromide, 1-bromo-2-phenoxyethane, benzene, 2-bromoethoxy, beta-phenoxyethyl bromide, 2-bromoethylphenylether, 1-2-bromoethoxy benzene | |
2-bromoethoxybenzene |
Specifications
589-10-6 | |
1.35 | |
65°C (149°F) | |
MFCD00000234 | |
2-bromoethoxy benzene, 2-bromoethyl phenyl ether, beta-bromophenetole, 2-phenoxyethylbromide, 2-phenoxyethyl bromide, 1-bromo-2-phenoxyethane, benzene, 2-bromoethoxy, beta-phenoxyethyl bromide, 2-bromoethylphenylether, 1-2-bromoethoxy benzene | |
JJFOBACUIRKUPN-UHFFFAOYSA-N | |
2-bromoethoxybenzene | |
68526 | |
98% |
31°C to 35°C | |
135°C to 137°C (20 mmHg) | |
C8H9BrO | |
508290 | |
Insoluble in water. Solubility in methanol is almost transparent. Soluble in chloroform and ethyl acetate. | |
C1=CC=C(C=C1)OCCBr | |
201.063 | |
201.07 | |
2-Phenoxyethyl bromide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
missing translation for 'einecsNumber' : 209-634-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 2-Phenoxyethyl bromide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : (2-bromoethoxy)benzene
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.