missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethanimidamide hydrochloride dihydrate, 97%, Thermo Scientific™

41.90 € - 119.00 €

Chemical Identifiers

CAS 67386-38-3
Molecular Formula C8H11ClN2O
Molecular Weight (g/mol) 186.64
MDL Number MFCD00173806
InChI Key ZLZLLGGMCNVVAS-UHFFFAOYSA-N
Synonym 2-phenoxyacetamidine hydrochloride, 2-phenoxyethanimidamide hydrochloride, 2-phenoxyacetimidamide hydrochloride, 2-phenoxy-acetamidine hcl, ethanimidamide, 2-phenoxy-, monohydrochloride, 2-phenoxyethanamidine, chloride, 2-phenoxyacetamidinehydrochloride, 2-phenoxy acetamidine hydrochloride, ethanimidamide,2-phenoxy-,hydrochloride 1:1
PubChem CID 2745912
IUPAC Name 2-phenoxyethanimidamide;hydrochloride
SMILES [H+].[Cl-].NC(=N)COC1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10464153
View Documents
Thermo Scientific Maybridge
MO07572DA
1 g
41.90 €
1g
Please to purchase this item. Need a web account? Register with us today!
10553603
View Documents
Thermo Scientific Maybridge
MO07572EA
10 g
119.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

67386-38-3
186.64
ZLZLLGGMCNVVAS-UHFFFAOYSA-N
2745912
[H+].[Cl-].NC(=N)COC1=CC=CC=C1
C8H11ClN2O
MFCD00173806
2-phenoxyacetamidine hydrochloride, 2-phenoxyethanimidamide hydrochloride, 2-phenoxyacetimidamide hydrochloride, 2-phenoxy-acetamidine hcl, ethanimidamide, 2-phenoxy-, monohydrochloride, 2-phenoxyethanamidine, chloride, 2-phenoxyacetamidinehydrochloride, 2-phenoxy acetamidine hydrochloride, ethanimidamide,2-phenoxy-,hydrochloride 1:1
2-phenoxyethanimidamide;hydrochloride

Specifications

67386-38-3
97%
C8H11ClN2O
2-phenoxyacetamidine hydrochloride, 2-phenoxyethanimidamide hydrochloride, 2-phenoxyacetimidamide hydrochloride, 2-phenoxy-acetamidine hcl, ethanimidamide, 2-phenoxy-, monohydrochloride, 2-phenoxyethanamidine, chloride, 2-phenoxyacetamidinehydrochloride, 2-phenoxy acetamidine hydrochloride, ethanimidamide,2-phenoxy-,hydrochloride 1:1
[H+].[Cl-].NC(=N)COC1=CC=CC=C1
186.64
222.68
2-Phenoxyethanimidamide hydrochloride dihydrate
97.0
Amber Glass Bottle
MFCD00173806
ZLZLLGGMCNVVAS-UHFFFAOYSA-N
2-phenoxyethanimidamide;hydrochloride
2745912
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.