Learn More
2-Nitrophenol, 98%, Thermo Scientific Chemicals
33.67 € - 313.56 €
Chemical Identifiers
CAS | 88-75-5 |
---|---|
Molecular Formula | C6H5NO3 |
Molecular Weight (g/mol) | 139.11 |
MDL Number | MFCD00011688 |
InChI Key | IQUPABOKLQSFBK-UHFFFAOYSA-N |
Synonym | o-nitrophenol, 2-hydroxynitrobenzene, phenol, 2-nitro, o-hydroxynitrobenzene, phenol, o-nitro, nitrophenol, o-nitrofenol, ortho-nitrophenol, phenol, nitro, o-nitrofenol czech |
PubChem CID | 6947 |
ChEBI | CHEBI:16260 |
IUPAC Name | 2-nitrophenol |
SMILES | OC1=CC=CC=C1[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11499443
|
Thermo Scientific Alfa Aesar
A12333.30 |
250 g |
33.67 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11489443
|
Thermo Scientific Alfa Aesar
A12333.0B |
1000 g |
82.51 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409453
|
Thermo Scientific Alfa Aesar
A12333.0I |
5000 g |
313.56 €
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Nitrophenol is a building block used as an acid-base indicator and corrosion inhibitor for aluminum-copper alloy. It is also used to make dyes, paint coloring, rubber chemicals, and substances that kill molds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Nitrophenol is a building block used as an acid-base indicator and corrosion inhibitor for aluminum-copper alloy. It is also used to make dyes, paint coloring, rubber chemicals, and substances that kill molds.
Solubility
Soluble in water (2.1 g/L at 20°C), ethanol, chloroform, alcohol, and acetone.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
Chemical Identifiers
88-75-5 | |
139.11 | |
IQUPABOKLQSFBK-UHFFFAOYSA-N | |
6947 | |
2-nitrophenol |
C6H5NO3 | |
MFCD00011688 | |
o-nitrophenol, 2-hydroxynitrobenzene, phenol, 2-nitro, o-hydroxynitrobenzene, phenol, o-nitro, nitrophenol, o-nitrofenol, ortho-nitrophenol, phenol, nitro, o-nitrofenol czech | |
CHEBI:16260 | |
OC1=CC=CC=C1[N+]([O-])=O |
Specifications
88-75-5 | |
1.26 | |
108°C (226°F) | |
C6H5NO3 | |
UN1663 | |
14,6620 | |
Soluble in water (2.1g/L at 20°C),ethanol,chloroform,alcohol,and acetone. | |
OC1=CC=CC=C1[N+]([O-])=O | |
139.11 | |
CHEBI:16260 | |
98% |
44°C to 48°C | |
214°C to 216°C | |
Phenol-like | |
MFCD00011688 | |
775403 | |
o-nitrophenol, 2-hydroxynitrobenzene, phenol, 2-nitro, o-hydroxynitrobenzene, phenol, o-nitro, nitrophenol, o-nitrofenol, ortho-nitrophenol, phenol, nitro, o-nitrofenol czech | |
IQUPABOKLQSFBK-UHFFFAOYSA-N | |
2-nitrophenol | |
6947 | |
139.11 | |
2-Nitrophenol |
Safety and Handling
GHS H Statement
H373-H302
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROPHENOLS
missing translation for 'einecsNumber' : 201-857-5
missing translation for 'rtecsNumber' : SM2100000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 2-Nitrophenol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : o-Nitrophenol
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.