missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nitro-1-propanol, 97%, Thermo Scientific Chemicals
50.10 € - 190.00 €
Chemical Identifiers
CAS | 2902-96-7 |
---|---|
Molecular Formula | C3H7NO3 |
Molecular Weight (g/mol) | 105.09 |
MDL Number | MFCD00007398 |
InChI Key | PCNWBUOSTLGPMI-UHFFFAOYNA-N |
Synonym | 2-nitro-1-propanol, 2-nitropropanol, 1-propanol, 2-nitro, 2-nitropropanol-1, ccris 5045, 2-nitro-propan-1-ol, acmc-20anp9, 1-hydroxy-2-nitropropane, 1-propanol, 2-nitro-8ci |
PubChem CID | 92187 |
IUPAC Name | 2-nitropropan-1-ol |
SMILES | CC(CO)[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11300508
|
Thermo Scientific Alfa Aesar
043843.06 |
5 g | |||||||
11310508
|
Thermo Scientific Alfa Aesar
043843.14 |
25 g |
190.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Nitro-1-propanol is used in improved synthesis of 2,2,2-trinitroethyl ethers, inhibits methane production during in vitro ruminal fermentation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2902-96-7 | |
105.09 | |
PCNWBUOSTLGPMI-UHFFFAOYNA-N | |
92187 | |
CC(CO)[N+]([O-])=O |
C3H7NO3 | |
MFCD00007398 | |
2-nitro-1-propanol, 2-nitropropanol, 1-propanol, 2-nitro, 2-nitropropanol-1, ccris 5045, 2-nitro-propan-1-ol, acmc-20anp9, 1-hydroxy-2-nitropropane, 1-propanol, 2-nitro-8ci | |
2-nitropropan-1-ol |
Specifications
2902-96-7 | |
73°C (1 mmHg) | |
C3H7NO3 | |
MFCD00007398 | |
Slightly soluble in water. | |
CC(CO)[N+]([O-])=O | |
105.09 | |
105.09 | |
Liquid |
1.185 g/mL | |
100°C (212°F) | |
1.439 | |
2-nitro-1-propanol, 2-nitropropanol, 1-propanol, 2-nitro, 2-nitropropanol-1, ccris 5045, 2-nitro-propan-1-ol, acmc-20anp9, 1-hydroxy-2-nitropropane, 1-propanol, 2-nitro-8ci | |
PCNWBUOSTLGPMI-UHFFFAOYNA-N | |
2-nitropropan-1-ol | |
92187 | |
98% | |
2-Nitro-1-propanol |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only