missing translation for 'onlineSavingsMsg'
Learn More

2-Methyltetrahydrofuran, 99+%, pure, stabilized, Thermo Scientific Chemicals

41.61 € - 226.13 €

Chemical Identifiers

CAS 96-47-9
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
InChI Key JWUJQDFVADABEY-UHFFFAOYSA-N
Synonym 2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran
PubChem CID 7301
IUPAC Name 2-methyloxolane
SMILES CC1CCCO1
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10387820
View Documents
Thermo Scientific Acros
168361000
100 g Glass bottle
41.61 €
100g
Please to purchase this item. Need a web account? Register with us today!
10082050
View Documents
Thermo Scientific Acros
168365000
500 g Glass bottle
113.57 €
500g
Please to purchase this item. Need a web account? Register with us today!
10713324
View Documents
Thermo Scientific Acros
168360010
1 kg Glass bottle
222.11 €
1kg
Please to purchase this item. Need a web account? Register with us today!
10328493
View Documents
Thermo Scientific Acros
168360025
2.5 kg Metal drum
226.13 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

96-47-9
86.13
2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran
2-methyloxolane
C5H10O
JWUJQDFVADABEY-UHFFFAOYSA-N
7301
CC1CCCO1

Specifications

-136.0°C
Colorless
78.0°C to 80.0°C
Liquid
99% min. (GC)
C5H10O
17,12
2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran
JWUJQDFVADABEY-UHFFFAOYSA-N
2-methyloxolane
7301
99+%
2-Methyltetrahydrofuran
96-47-9
0.8600g/mL
Authentic
1.4046 to 1.4066
0.86
4 mPa.s (25°C)
15,6198
Solubility in water: 150g/l (25°C)
CC1CCCO1
86.13
86.13
Pure
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May form explosive peroxides.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 202-507-4

Product Identifier
  • 2-Methyltetrahydrofuran

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.