missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylamino)pyrimidine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Product Code. 15461476
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-(Methylamino)pyrimidine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylamino)pyrimidine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 904326-88-1
Molecular Formula C11H18BN3O2
Molecular Weight (g/mol) 235.094
MDL Number MFCD11878345
InChI Key QDOXNCAIXITTKA-UHFFFAOYSA-N
Synonym n-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-methylaminopyrimidine-5-boronic acid, pinacol ester, n-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-methylaminopyrimidine-5-boronic acid,pinacol ester, 2-methylamino pyrimidine-5-boronic acid pinacol ester, 2-methylamino pyrimidin-5-yl boronic acid pinacol ester, methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrimidin-2-yl-amine, methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrimidin-2-yl amine, n-m ethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, n-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyrimidin-2-amine
PubChem CID 46739641
IUPAC Name N-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2)NC

Specifications

Melting Point 122°C to 125°C
Quantity 1 g
Formula Weight 235.09
Percent Purity 96%
Chemical Name or Material 2-(Methylamino)pyrimidine-5-boronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'