missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-(trifluoromethoxy)acetophenone, 97%, Thermo Scientific™

Product Code. 15446277
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1373920-71-8
Molecular Formula C10H9F3O2
Molecular Weight (g/mol) 218.175
MDL Number MFCD22201058
InChI Key XVSBANNQMCDRTC-UHFFFAOYSA-N
Synonym 2'-methyl-5'-trifluoromethoxy acetophenone, 1-2-methyl-5-trifluoromethoxy phenyl ethanone, 2-methyl-5-trifluoromethoxy acetophenone
PubChem CID 86277664
IUPAC Name 1-[2-methyl-5-(trifluoromethoxy)phenyl]ethanone
SMILES CC1=C(C=C(C=C1)OC(F)(F)F)C(=O)C

Specifications

Quantity 1 g
Formula Weight 218.17
Percent Purity 97%
Chemical Name or Material 2-Methyl-5-(trifluoromethoxy)acetophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'