Learn More
2-(Methyl-2-pyridylamino)ethanol, 98%, Thermo Scientific Chemicals
68.20 € - 251.00 €
Chemical Identifiers
CAS | 122321-04-4 |
---|---|
Molecular Formula | C8H12N2O |
Molecular Weight (g/mol) | 152.197 |
MDL Number | MFCD02667633 |
InChI Key | MWGKOPUDDQZERY-UHFFFAOYSA-N |
Synonym | 2-methyl pyridin-2-yl amino ethanol, 2-methyl-2-pyridylamino ethanol, 2-methyl pyridin-2-yl amino ethan-1-ol, 2-n-methyl-2-pyridylaminoethanol, 2-methyl-2-pyridinylamino ethanol, 2-n-methyl-2-pyridylamino ethanol, 2-n-methyl-n-2-pyridinyl amino ethanol, 2-n-2-hydroxyethyl-n-methylamino pyridine, 2-methyl 2-pyridinyl amino-1-ethanol |
PubChem CID | 737420 |
IUPAC Name | 2-[methyl(pyridin-2-yl)amino]ethanol |
SMILES | CN(CCO)C1=CC=CC=N1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11467236
|
Thermo Scientific Alfa Aesar
B25555.18 |
50 g |
68.20 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11477236
|
Thermo Scientific Alfa Aesar
B25555.30 |
250 g | |||||||
Description
2-Methyl-2-pyridylaminoethanol is used as a reagent to synthesize Rosiglitazone (Maleate: R693500), a thiazolinedione drug used to reduce insulin resistance in patients with Type 2 diabetes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methyl-2-pyridylaminoethanol is used as a reagent to synthesize Rosiglitazone (Maleate: R693500), a thiazolinedione drug used to reduce insulin resistance in patients with Type 2 diabetes.
Solubility
Soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
122321-04-4 | |
152.197 | |
MWGKOPUDDQZERY-UHFFFAOYSA-N | |
737420 | |
CN(CCO)C1=CC=CC=N1 |
C8H12N2O | |
MFCD02667633 | |
2-methyl pyridin-2-yl amino ethanol, 2-methyl-2-pyridylamino ethanol, 2-methyl pyridin-2-yl amino ethan-1-ol, 2-n-methyl-2-pyridylaminoethanol, 2-methyl-2-pyridinylamino ethanol, 2-n-methyl-2-pyridylamino ethanol, 2-n-methyl-n-2-pyridinyl amino ethanol, 2-n-2-hydroxyethyl-n-methylamino pyridine, 2-methyl 2-pyridinyl amino-1-ethanol | |
2-[methyl(pyridin-2-yl)amino]ethanol |
Specifications
122321-04-4 | |
1.575 | |
2-methyl pyridin-2-yl amino ethanol, 2-methyl-2-pyridylamino ethanol, 2-methyl pyridin-2-yl amino ethan-1-ol, 2-n-methyl-2-pyridylaminoethanol, 2-methyl-2-pyridinylamino ethanol, 2-n-methyl-2-pyridylamino ethanol, 2-n-methyl-n-2-pyridinyl amino ethanol, 2-n-2-hydroxyethyl-n-methylamino pyridine, 2-methyl 2-pyridinyl amino-1-ethanol | |
MWGKOPUDDQZERY-UHFFFAOYSA-N | |
2-[methyl(pyridin-2-yl)amino]ethanol | |
737420 | |
98% |
C8H12N2O | |
MFCD02667633 | |
Soluble in water. | |
CN(CCO)C1=CC=CC=N1 | |
152.197 | |
152.19 | |
2-(Methyl-2-pyridylamino)ethanol |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only