missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Methyl-1,3-thiazol-4-yl)methanol, 97%, Thermo Scientific™
124.00 € - 190.00 €
Chemical Identifiers
CAS | 76632-23-0 |
---|---|
Molecular Formula | C5H7NOS |
Molecular Weight (g/mol) | 129.177 |
InChI Key | KRCIFTBSQKDYMH-UHFFFAOYSA-N |
Synonym | 2-methyl-1,3-thiazol-4-yl methanol, 2-methylthiazol-4-yl methanol, 4-hydroxymethyl-2-methylthiazole, 4-thiazolemethanol, 2-methyl, 2-methyl-thiazol-4-yl-methanol, 2-methyl-4-thiazolemethanol, 4-thiazolemethanol,2-methyl, 2-methyl-4-thiazolyl methanol, 4-hydroxymethyl-2-methyl thiazole |
PubChem CID | 10582809 |
IUPAC Name | (2-methyl-1,3-thiazol-4-yl)methanol |
SMILES | CC1=NC(=CS1)CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10603595
|
Thermo Scientific Maybridge
CC05709DA |
1 g | Amber Glass Bottle |
124.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10006924
|
Thermo Scientific Maybridge
CC05709DE |
5 g | Amber Glass Bottle |
190.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
76632-23-0 | |
129.177 | |
2-methyl-1,3-thiazol-4-yl methanol, 2-methylthiazol-4-yl methanol, 4-hydroxymethyl-2-methylthiazole, 4-thiazolemethanol, 2-methyl, 2-methyl-thiazol-4-yl-methanol, 2-methyl-4-thiazolemethanol, 4-thiazolemethanol,2-methyl, 2-methyl-4-thiazolyl methanol, 4-hydroxymethyl-2-methyl thiazole | |
(2-methyl-1,3-thiazol-4-yl)methanol |
C5H7NOS | |
KRCIFTBSQKDYMH-UHFFFAOYSA-N | |
10582809 | |
CC1=NC(=CS1)CO |
Specifications
76632-23-0 | |
C5H7NOS | |
KRCIFTBSQKDYMH-UHFFFAOYSA-N | |
(2-methyl-1,3-thiazol-4-yl)methanol | |
10582809 | |
97% |
97% | |
2-methyl-1,3-thiazol-4-yl methanol, 2-methylthiazol-4-yl methanol, 4-hydroxymethyl-2-methylthiazole, 4-thiazolemethanol, 2-methyl, 2-methyl-thiazol-4-yl-methanol, 2-methyl-4-thiazolemethanol, 4-thiazolemethanol,2-methyl, 2-methyl-4-thiazolyl methanol, 4-hydroxymethyl-2-methyl thiazole | |
CC1=NC(=CS1)CO | |
129.177 | |
129.18 | |
(2-Methyl-1, 3-thiazol-4-yl)methanol |
Safety and Handling
GHS Signal Word: Warning
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
(2-Methyl-1,3-thiazol-4-yl)methanol, 97%, Thermo Scientific™