Learn More
2-Mesitylenesulfonyl chloride, 99%, Thermo Scientific Chemicals
45.43 € - 488.43 €
Chemical Identifiers
CAS | 773-64-8 |
---|---|
Molecular Formula | C9H11ClO2S |
Molecular Weight (g/mol) | 218.7 |
MDL Number | MFCD00007434 |
InChI Key | PVJZBZSCGJAWNG-UHFFFAOYSA-N |
Synonym | 2-mesitylenesulfonyl chloride, mesitylenesulfonyl chloride, 2,4,6-trimethylbenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4,6-trimethyl, mesitylsulfonyl chloride, 2-mesitylenesulfonylchloride, mesitylene-2-sulphonyl chloride, 2,4,6-trimethyl-benzenesulfonyl chloride, 2,4,6-trimethylphenylsulfonyl chloride, mesitylsulfonylchloride |
PubChem CID | 13046 |
IUPAC Name | 2,4,6-trimethylbenzenesulfonyl chloride |
SMILES | CC1=CC(=C(C(=C1)C)S(=O)(=O)Cl)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10081270
|
Thermo Scientific Acros
125590250 |
25 g | Plastic bottle |
45.43 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10376810
|
Thermo Scientific Acros
125591000 |
100 g | Plastic bottle |
118.59 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10406750
|
Thermo Scientific Acros
125595000 |
500 g | Plastic bottle |
488.43 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
773-64-8 | |
218.7 | |
PVJZBZSCGJAWNG-UHFFFAOYSA-N | |
13046 | |
CC1=CC(=C(C(=C1)C)S(=O)(=O)Cl)C |
C9H11ClO2S | |
MFCD00007434 | |
2-mesitylenesulfonyl chloride, mesitylenesulfonyl chloride, 2,4,6-trimethylbenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4,6-trimethyl, mesitylsulfonyl chloride, 2-mesitylenesulfonylchloride, mesitylene-2-sulphonyl chloride, 2,4,6-trimethyl-benzenesulfonyl chloride, 2,4,6-trimethylphenylsulfonyl chloride, mesitylsulfonylchloride | |
2,4,6-trimethylbenzenesulfonyl chloride |
Specifications
773-64-8 | |
100.0 | |
Gray to White | |
>110°C | |
98.5% min. (ex Cl) (Argentometry) | |
(CH3)3C6H2SO2Cl | |
11, 136 | |
2-mesitylenesulfonyl chloride, mesitylenesulfonyl chloride, 2,4,6-trimethylbenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4,6-trimethyl, mesitylsulfonyl chloride, 2-mesitylenesulfonylchloride, mesitylene-2-sulphonyl chloride, 2,4,6-trimethyl-benzenesulfonyl chloride, 2,4,6-trimethylphenylsulfonyl chloride, mesitylsulfonylchloride | |
PVJZBZSCGJAWNG-UHFFFAOYSA-N | |
2,4,6-trimethylbenzenesulfonyl chloride | |
13046 | |
99% | |
2-Mesitylenesulfonyl chloride, 99% |
98.5 | |
53°C to 57°C | |
150°C (20.0 mmHg) | |
Authentic | |
C9H11ClO2S | |
MFCD00007434 | |
01,661; 05,430; 06,625; 08,318 | |
Solubility in water: reacts. Other solubilities: soluble in ether,toluene and dichloromethane | |
CC1=CC(=C(C(=C1)C)S(=O)(=O)Cl)C | |
218.7 | |
218.7 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 212-257-8
missing translation for 'rtecsNumber' : DB8930000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.