missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-3-phenyl-3,5,6,7-tetrahydro-4H-cyclopenta[b]thieno[2,3-d]pyrimidin-4-one, 96%, Thermo Scientific™

Product Code. 15455175
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 300557-77-1
Molecular Formula C15H12N2OS2
Molecular Weight (g/mol) 300.39
MDL Number MFCD01018198
InChI Key LLFBYHAEKFMVIS-UHFFFAOYSA-N
Synonym 2-mercapto-3-phenyl-3,5,6,7-tetrahydro-4h-cyclopenta 4,5 thieno 2,3-d pyrimidin-4-one, 6-mercapto-5-phenyl-1,2,3,5-tetrahydro-8-thia-5,7-diaza-cyclopenta a inden-4-one, 2-mercapto-3-phenyl-6,7-dihydro-3h-cyclopenta 4,5 thieno 2,3-d pyrimidin-4 5h-one, 3-phenyl-2-sulfanyl-3,5,6,7-tetrahydro-4h-cyclopenta 4,5 thieno 2,3-d pyrimidin-4-one, 4h-cyclopenta 4,5 thieno 2,3-d pyrimidin-4-one,1,2,3,5,6,7-hexahydro-3-phenyl-2-thioxo, 11-phenyl-10-sulfanyl-7-thia-9,11-diazatricyclo 6.4.0.0 2 ,? dodeca-1 8 ,2 6 ,9-trien-12-one, 11-phenyl-10-sulfanylidene-7-thia-9,11-diazatricyclo 6.4.0.0 2 ,? dodeca-1 8 ,2 6-dien-12-one, cbmicro_017927, 1,2,3,4-tetrahydro-2-thioxo-3-phenyl-5,6-propanothieno 2,3-d pyrimidine-4-one, 1,2,6,7-tetrahydro-3-phenyl-2-thioxo-5h-cyclopenta 4,5 thieno 2,3-d pyrimidin-4 3h-one
PubChem CID 743434
SMILES O=C1N(C(=S)NC2=C1C1=C(CCC1)S2)C1=CC=CC=C1

Specifications

Melting Point >250°C
Quantity 5 g
IUPAC Name 2-phenyl-3-sulfanylidene-4,6,7,8-tetrahydrocyclopenta[2,3]thieno[2,4-b]pyrimidin-1-one
Formula Weight 300.4
Percent Purity 96%
Chemical Name or Material 2-Mercapto-3-phenyl-3,5,6,7-tetrahydro-4H-cyclopenta[b]thieno[2,3-d]pyrimidin-4-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'