missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Isopropoxybenzeneboronic acid, 95%, Thermo Scientific Chemicals
38.30 € - 136.00 €
Chemical Identifiers
CAS | 138008-97-6 |
---|---|
Molecular Formula | C9H13BO3 |
Molecular Weight (g/mol) | 180.01 |
MDL Number | MFCD03427050 |
InChI Key | XDMKBIIRBDPSOE-UHFFFAOYSA-N |
Synonym | 2-isopropoxyphenylboronic acid, 2-isopropoxyphenyl boronic acid, 2-isopoxybenzeneboronic acid, 2-isopropoxybenzeneboronic acid, boronic acid, 2-1-methylethoxy phenyl, 2-propan-2-yloxy phenylboronic acid, 2-isopropoxyphenylboronicacid, 2-propan-2-yloxyphenyl boronic acid, akos brn-0366, pubchem13297 |
PubChem CID | 5084099 |
IUPAC Name | (2-propan-2-yloxyphenyl)boronic acid |
SMILES | CC(C)OC1=CC=CC=C1B(O)O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
138008-97-6 | |
180.01 | |
XDMKBIIRBDPSOE-UHFFFAOYSA-N | |
5084099 | |
CC(C)OC1=CC=CC=C1B(O)O |
C9H13BO3 | |
MFCD03427050 | |
2-isopropoxyphenylboronic acid, 2-isopropoxyphenyl boronic acid, 2-isopoxybenzeneboronic acid, 2-isopropoxybenzeneboronic acid, boronic acid, 2-1-methylethoxy phenyl, 2-propan-2-yloxy phenylboronic acid, 2-isopropoxyphenylboronicacid, 2-propan-2-yloxyphenyl boronic acid, akos brn-0366, pubchem13297 | |
(2-propan-2-yloxyphenyl)boronic acid |
Specifications
138008-97-6 | |
1.075 | |
110°C (230°F) | |
1.509 | |
2-isopropoxyphenylboronic acid, 2-isopropoxyphenyl boronic acid, 2-isopoxybenzeneboronic acid, 2-isopropoxybenzeneboronic acid, boronic acid, 2-1-methylethoxy phenyl, 2-propan-2-yloxy phenylboronic acid, 2-isopropoxyphenylboronicacid, 2-propan-2-yloxyphenyl boronic acid, akos brn-0366, pubchem13297 | |
CC(C)OC1=CC=CC=C1B(O)O | |
180.01 | |
180.01 | |
2-Isopropoxybenzeneboronic acid |
30°C to 35°C | |
199°C | |
C9H13BO3 | |
MFCD03427050 | |
XDMKBIIRBDPSOE-UHFFFAOYSA-N | |
(2-propan-2-yloxyphenyl)boronic acid | |
5084099 | |
95% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only