missing translation for 'onlineSavingsMsg'
Learn More

2-Isocyanatobenzonitrile, 97%, Thermo Scientific™

Product Code. 10502632
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 42066-86-4
Molecular Formula C8H4N2O
Molecular Weight (g/mol) 144.133
InChI Key FQLNVWAFXQUXGW-UHFFFAOYSA-N
PubChem CID 2735873
IUPAC Name 2-isocyanatobenzonitrile
SMILES C1=CC=C(C(=C1)C#N)N=C=O

Specifications

Assay Percent Range 95%
Packaging Amber glass bottle
Linear Formula CNC6H4NCO
Quantity 1 g
Synonym 2-cyanophenyl isocyanate, benzonitrile,2-isocyanato, 2-isocyanato-benzonitrile, 2-isocyanatobenzenecarbonitrile, 2-cyanophenylisocyanate, benzonitrile, 2-isocyanato, acmc-1ap1b
Formula Weight 144.13
Percent Purity 95%
Chemical Name or Material 2-Isocyanatobenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'