Learn More
2-Iodophenol, 98%, Thermo Scientific Chemicals
23.82 € - 215.07 €
Chemical Identifiers
CAS | 533-58-4 |
---|---|
Molecular Formula | C6H5IO |
Molecular Weight (g/mol) | 220.01 |
MDL Number | MFCD00013963 |
InChI Key | KQDJTBPASNJQFQ-UHFFFAOYSA-N |
Synonym | o-iodophenol, phenol, 2-iodo, phenol, o-iodo, o-jodfenol, 2-jodfenol, phenol, iodo, o-jodfenol czech, 2-jodfenol czech, o-jodphenol, 2-iodanylphenol |
PubChem CID | 10784 |
ChEBI | CHEBI:16706 |
IUPAC Name | 2-iodophenol |
SMILES | OC1=CC=CC=C1I |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11470144
|
Thermo Scientific Alfa Aesar
A13599.06 |
5 g |
23.82 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480144
|
Thermo Scientific Alfa Aesar
A13599.14 |
25 g |
77.99 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11490144
|
Thermo Scientific Alfa Aesar
A13599.22 |
100 g |
215.07 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Iodophenol is used in the synthesis of aryl 2-benzofuranyl and aryl 2-indolyl carbinols of high enantiomeric purity and 3,3-disubstituted-2,3-dihydrobenzofurans. Coupling with phenylacetylene in the presence of a Pd complex and CuI gives 2-phenylbenzofuran in good yield.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Iodophenol is used in the synthesis of aryl 2-benzofuranyl and aryl 2-indolyl carbinols of high enantiomeric purity and 3,3-disubstituted-2,3-dihydrobenzofurans. Coupling with phenylacetylene in the presence of a Pd complex and CuI gives 2-phenylbenzofuran in good yield.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to light. Incompatible with oxidizing agents.
Chemical Identifiers
533-58-4 | |
220.01 | |
KQDJTBPASNJQFQ-UHFFFAOYSA-N | |
10784 | |
2-iodophenol |
C6H5IO | |
MFCD00013963 | |
o-iodophenol, phenol, 2-iodo, phenol, o-iodo, o-jodfenol, 2-jodfenol, phenol, iodo, o-jodfenol czech, 2-jodfenol czech, o-jodphenol, 2-iodanylphenol | |
CHEBI:16706 | |
OC1=CC=CC=C1I |
Specifications
533-58-4 | |
1.947 | |
>110°C (230°F) | |
MFCD00013963 | |
Light sensitive | |
o-iodophenol, phenol, 2-iodo, phenol, o-iodo, o-jodfenol, 2-jodfenol, phenol, iodo, o-jodfenol czech, 2-jodfenol czech, o-jodphenol, 2-iodanylphenol | |
KQDJTBPASNJQFQ-UHFFFAOYSA-N | |
2-iodophenol | |
10784 | |
220.01 | |
2-Iodophenol |
39°C to 41°C | |
186°C to 187°C (160 mmHg) | |
C6H5IO | |
1855300 | |
14,5035 | |
Soluble in water. | |
OC1=CC=CC=C1I | |
220.01 | |
CHEBI:16706 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'einecsNumber' : 208-569-9
missing translation for 'rtecsNumber' : SL5500000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only