missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 2-Hydroxy-5-(trifluoromethoxy)benzonitrile, 95%
74.00 € - 205.00 €
Chemical Identifiers
CAS | 875664-40-7 |
---|---|
Molecular Formula | C8H4F3NO2 |
Molecular Weight (g/mol) | 203.12 |
MDL Number | MFCD04973781 |
InChI Key | GOAVRYAOBXORSY-UHFFFAOYSA-N |
Synonym | 2-hydroxy-5-trifluoromethoxy benzonitrile |
PubChem CID | 24721643 |
IUPAC Name | 2-hydroxy-5-(trifluoromethoxy)benzonitrile |
SMILES | C1=CC(=C(C=C1OC(F)(F)F)C#N)O |
Description
Chemical Identifiers
875664-40-7 | |
203.12 | |
GOAVRYAOBXORSY-UHFFFAOYSA-N | |
24721643 | |
C1=CC(=C(C=C1OC(F)(F)F)C#N)O |
C8H4F3NO2 | |
MFCD04973781 | |
2-hydroxy-5-trifluoromethoxy benzonitrile | |
2-hydroxy-5-(trifluoromethoxy)benzonitrile |
Specifications
875664-40-7 | |
C8H4F3NO2 | |
UN3439 | |
GOAVRYAOBXORSY-UHFFFAOYSA-N | |
2-hydroxy-5-(trifluoromethoxy)benzonitrile | |
24721643 | |
95% |
84°C to 90°C | |
MFCD04973781 | |
2-hydroxy-5-trifluoromethoxy benzonitrile | |
C1=CC(=C(C=C1OC(F)(F)F)C#N)O | |
203.12 | |
203.12 | |
2-Hydroxy-5-(trifluoromethoxy)benzonitrile |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures