missing translation for 'onlineSavingsMsg'
Learn More

2-Hexynoic acid, 96%, Thermo Scientific Chemicals

26.60 € - 73.00 €

Chemical Identifiers

CAS 764-33-0
Molecular Weight (g/mol) 112.128
MDL Number MFCD00014371
InChI Key AKYAUBWOTZJUBI-UHFFFAOYSA-N
Synonym 2-hexynoic acid, hexynoic acid, acmc-1bezk, akyaubwotzjubi-uhfffaoysa
PubChem CID 324380
IUPAC Name hex-2-ynoic acid
SMILES CCCC#CC(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11357735
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L02526.03
1 g
26.60 €
1g
Please to purchase this item. Need a web account? Register with us today!
11367735
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L02526.06
5 g
73.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

764-33-0
112.128
AKYAUBWOTZJUBI-UHFFFAOYSA-N
324380
CCCC#CC(=O)O
C6H8O2
MFCD00014371
2-hexynoic acid, hexynoic acid, acmc-1bezk, akyaubwotzjubi-uhfffaoysa
hex-2-ynoic acid

Specifications

764-33-0
110°C to 112°C (10 mmHg)
1.459
UN3261
2-hexynoic acid, hexynoic acid, acmc-1bezk, akyaubwotzjubi-uhfffaoysa
CCCC#CC(=O)O
112.128
112.13
2-Hexynoic acid
23°C to 26°C
C6H8O2
MFCD00014371
1744983
AKYAUBWOTZJUBI-UHFFFAOYSA-N
hex-2-ynoic acid
324380
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only