missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Heptanone, 99%, Honeywell™
0.99
32.67 € - 181.45 €
Chemical Identifiers
CAS | 110-43-0 |
---|---|
Molecular Formula | C7H14O |
Molecular Weight (g/mol) | 114.19 |
MDL Number | MFCD00009513 |
InChI Key | CATSNJVOTSVZJV-UHFFFAOYSA-N |
Synonym | 2-heptanone, methyl amyl ketone, butylacetone, methyl pentyl ketone, methyl n-amyl ketone, amyl methyl ketone, n-amyl methyl ketone, n-pentyl methyl ketone, heptanone, pentyl methyl ketone |
PubChem CID | 8051 |
ChEBI | CHEBI:5672 |
IUPAC Name | heptan-2-one |
SMILES | CCCCCC(C)=O |
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Chemical Identifiers
110-43-0 | |
114.19 | |
CATSNJVOTSVZJV-UHFFFAOYSA-N | |
8051 | |
heptan-2-one |
C7H14O | |
MFCD00009513 | |
2-heptanone, methyl amyl ketone, butylacetone, methyl pentyl ketone, methyl n-amyl ketone, amyl methyl ketone, n-amyl methyl ketone, n-pentyl methyl ketone, heptanone, pentyl methyl ketone | |
CHEBI:5672 | |
CCCCCC(C)=O |
Specifications
110-43-0 | |
C7H14O | |
MFCD00009513 | |
1699063 | |
CATSNJVOTSVZJV-UHFFFAOYSA-N | |
heptan-2-one | |
8051 | |
114.19g/mol | |
2-Heptanone |
0.82g/mL at 25°C | |
CH3(CH2)4COCH3 | |
UN1110 | |
2-heptanone, methyl amyl ketone, butylacetone, methyl pentyl ketone, methyl n-amyl ketone, amyl methyl ketone, n-amyl methyl ketone, n-pentyl methyl ketone, heptanone, pentyl methyl ketone | |
CCCCCC(C)=O | |
114.19 | |
CHEBI:5672 | |
99% |
Safety and Handling
missing translation for 'dotInformation' : class 3 - PG 3 - n-Amyl methyl ketone
missing translation for 'rtecsNumber' : 203-767-1
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title