missing translation for 'onlineSavingsMsg'
Learn More

2-Furoyl chloride, 98+%, Thermo Scientific Chemicals

Product Code. 11307076
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 527-69-5
Molecular Formula C5H3ClO2
Molecular Weight (g/mol) 130.527
MDL Number MFCD00003228
InChI Key OFTKFKYVSBNYEC-UHFFFAOYSA-N
Synonym 2-furoyl chloride, furoyl chloride, 2-furancarbonyl chloride, furancarbonyl chloride, 2-furoylchloride, unii-2us0dxi75s, 2us0dxi75s, 2-furancarboxylic acid chloride, furan-2-carboxylic acid chloride, furanoyl chloride
PubChem CID 68242
IUPAC Name furan-2-carbonyl chloride
SMILES C1=COC(=C1)C(=O)Cl

Specifications

Melting Point -2°C to +1°C
Density 1.325
Boiling Point 173°C to 174°C
Flash Point 85°C (185°F)
Refractive Index 1.531
Quantity 250 g
UN Number UN3265
Beilstein 110144
Sensitivity Moisture sensitive
Merck Index 14,4310
Formula Weight 130.53
Percent Purity ≥98%
Chemical Name or Material 2-Furoyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'