missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-N-(4-fluorobenzyl)aniline, 97%, Thermo Scientific™

Chemical Identifiers

CAS 886791-73-7
Molecular Formula C13H11F2N
Molecular Weight (g/mol) 219.235
InChI Key KQBMUYYUEWLGQP-UHFFFAOYSA-N
Synonym 2-fluoro-n-4-fluorobenzyl aniline, 2-fluoro-n-4-fluorophenyl methyl aniline, 4-fluorobenzylamine, n-2-fluorophenyl
PubChem CID 6422135
IUPAC Name 2-fluoro-N-[(4-fluorophenyl)methyl]aniline
SMILES C1=CC=C(C(=C1)NCC2=CC=C(C=C2)F)F
View More Specs

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
A novel class of potent CCR3 receptor antagonists were designed and synthesized from the derivative of this compound.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Chemical Identifiers

886791-73-7
219.235
2-fluoro-n-4-fluorobenzyl aniline, 2-fluoro-n-4-fluorophenyl methyl aniline, 4-fluorobenzylamine, n-2-fluorophenyl
2-fluoro-N-[(4-fluorophenyl)methyl]aniline
C13H11F2N
KQBMUYYUEWLGQP-UHFFFAOYSA-N
6422135
C1=CC=C(C(=C1)NCC2=CC=C(C=C2)F)F

Specifications

886791-73-7
2-fluoro-n-4-fluorobenzyl aniline, 2-fluoro-n-4-fluorophenyl methyl aniline, 4-fluorobenzylamine, n-2-fluorophenyl
C1=CC=C(C(=C1)NCC2=CC=C(C=C2)F)F
219.235
219.23
2-Fluoro-N-(4-fluorobenzyl)aniline
C13H11F2N
KQBMUYYUEWLGQP-UHFFFAOYSA-N
2-fluoro-N-[(4-fluorophenyl)methyl]aniline
6422135
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications