missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethoxy)-DL-phenylalanine, 97%, Thermo Scientific™

Product Code. 15428405
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1391008-15-3
Molecular Formula C10H9F4NO3
Molecular Weight (g/mol) 267.18
MDL Number MFCD16652500
InChI Key POFOCSWADMUTGH-UHFFFAOYSA-N
Synonym 2-fluoro-4-trifluoromethoxy-dl-phenylalanine, 2-amino-3-2-fluoro-4-trifluoromethoxy phenyl propanoic acid
PubChem CID 66523488
IUPAC Name 2-amino-3-[2-fluoro-4-(trifluoromethoxy)phenyl]propanoic acid
SMILES C1=CC(=C(C=C1OC(F)(F)F)F)CC(C(=O)O)N

Specifications

Formula Weight 267.18
Percent Purity 97%
Quantity 1 g
Chemical Name or Material 2-Fluoro-4-(trifluoromethoxy)-DL-phenylalanine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'