missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Fluoro-4'-methoxyacetophenone, 99%, Thermo Scientific™
36.70 € - 449.00 €
Chemical Identifiers
CAS | 74457-86-6 |
---|---|
Molecular Formula | C9H9FO2 |
Molecular Weight (g/mol) | 168.17 |
MDL Number | MFCD00042290 |
InChI Key | PIRRWUMTIBFCCW-UHFFFAOYSA-N |
Synonym | 2'-fluoro-4'-methoxyacetophenone, 2-fluoro-4-methoxyacetophenone, 1-2-fluoro-4-methoxyphenyl ethanone, 1-2-fluoro-4-methoxyphenyl ethan-1-one, 4-acetyl-3-fluoroanisole, 1-acetyl-2-fluoro-4-methoxybenzene, 2'-fluor-4'-methoxyacetophenone, 2-fluoro-4-methoxy-acetophenone, ethanone, 1-2-fluoro-4-methoxyphenyl, zlchem 344 |
PubChem CID | 592821 |
IUPAC Name | 1-(2-fluoro-4-methoxyphenyl)ethanone |
SMILES | COC1=CC=C(C(C)=O)C(F)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15127422
|
Thermo Scientific Acros
263540050 |
5 g | Glass bottle | ||||||
15147422
|
Thermo Scientific Acros
263540250 |
25 g | Glass bottle | ||||||
15157422
|
Thermo Scientific Acros
263541000 |
100 g | Glass bottle | ||||||
Description
Chemical Identifiers
74457-86-6 | |
168.17 | |
PIRRWUMTIBFCCW-UHFFFAOYSA-N | |
592821 | |
COC1=CC=C(C(C)=O)C(F)=C1 |
C9H9FO2 | |
MFCD00042290 | |
2'-fluoro-4'-methoxyacetophenone, 2-fluoro-4-methoxyacetophenone, 1-2-fluoro-4-methoxyphenyl ethanone, 1-2-fluoro-4-methoxyphenyl ethan-1-one, 4-acetyl-3-fluoroanisole, 1-acetyl-2-fluoro-4-methoxybenzene, 2'-fluor-4'-methoxyacetophenone, 2-fluoro-4-methoxy-acetophenone, ethanone, 1-2-fluoro-4-methoxyphenyl, zlchem 344 | |
1-(2-fluoro-4-methoxyphenyl)ethanone |
Specifications
74457-86-6 | |
100.0 | |
Colorless | |
>110°C | |
98.5% min. (GC) | |
MFCD00042290 | |
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents | |
COC1=CC=C(C(C)=O)C(F)=C1 | |
168.17 | |
168.17 | |
Viscous Solution |
98.5 | |
50.0°C to 54.0°C | |
80.0°C to 85.0°C (0.1 mmHg) | |
Authentic | |
C9H9FO2 | |
2'-fluoro-4'-methoxyacetophenone, 2-fluoro-4-methoxyacetophenone, 1-2-fluoro-4-methoxyphenyl ethanone, 1-2-fluoro-4-methoxyphenyl ethan-1-one, 4-acetyl-3-fluoroanisole, 1-acetyl-2-fluoro-4-methoxybenzene, 2'-fluor-4'-methoxyacetophenone, 2-fluoro-4-methoxy-acetophenone, ethanone, 1-2-fluoro-4-methoxyphenyl, zlchem 344 | |
PIRRWUMTIBFCCW-UHFFFAOYSA-N | |
1-(2-fluoro-4-methoxyphenyl)ethanone | |
592821 | |
99% | |
2'-Fluoro-4'-methoxyacetophenone, 99% |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'tsca' : TSCA
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2'-Fluoro-4'-methoxyacetophenone
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only